Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X)
Cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines.
Brand:  Invitrogen™ 00-4970-03
Additional Details : Weight : 1.00000kg Transport : UN number : 1170 Chem class : 3 Pack group : II
 View more versions of this product
Product Code. 15290035
  • 2700.00 SEK / 400µL
In Stock
to see stock.
Description
Description
  • Can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays
  • Can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression
Specifications
Specifications
4 x 100μL
Cell Stimulation Cocktail
eBioscience™
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • eBioscience Cell Stimulation Cocktail (500X)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Certificates
Ofertas especiales
Ofertas especiales
For Research Use Only. Not for use in diagnostic procedures.