Läs mer
Greiner Bio-One CELLSTAR Cellkulturflaskor
Beskrivning
- Improved cell adhesion through physical surface treatment
- Neck of high profile flask reduces the risk of wetting the inside of the cap with medium; provides additional protection against contamination
- Canted neck
- TC surface treatment
- Graduation on the side
- Stackable
- Measuring grid
- Red standard screw cap
- Sterile and user-friendly packaging
- Free of detectable DNase, RNase, human DNA, non-pyrogenic
- Non-Cytotoxic
Specifikationer
Specifikationer
| Stängningsfärg | Red |
| Stängningsmaterial | HDPE |
| Kapacitet (metrisk) | 50 mL |
| Material | Polystyrene |
| Inkluderar | Printed and engraved graduations |
| Hals stil | Canted |
| Ytbeläggning | Physical Surface Treatment |
| Graderad | Yes |
| Sterilitet | Sterile |
| Förslutningstyp | Filter Screw Cap |
| Visa mer |