missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cellulose Acetate 2-D Gel Electrophoresis System

Full-featured two-dimensional gel electrophoresis system. Fisherbrand™ Cellulose Acetate 2-D Gel Electrophoresis System accommodates up to three standard membranes simultaneously.

Brand:  Fisherbrand™

Additional Details : Weight : 0.01000kg

Product Code. 12395488

  • 9540.00 SEK / Each


Estimated Shipment
07-10-2020
Add to basket

Description

Description

  • Asymmetric offset electrodes ensures that membrane is run in correct polar orientation
  • Clamping bars within clip holders (holds cellulose acetate membrane or paper wick gently in position)
  • Adjustable bridge prices to support cellulose acetate membranes
  • Central partition to separate buffer
  • Active tank area: 24.5 x 26cm (W x L), accomodates 145 x 192mm membranes or three standard 78 x 75/150mm membranes simultaneously

Applications:

  • Two-dimensional screening of inherited metabolic disorders in children
Specifications

Specifications

300 x 320 x 65 mm
SDS PAGE
Cellulose Acetate
2-D Electrophoresis Gel System
4mm power output connectors
1200 mL
Videos
Documents