Learn More
Abnova™ CEN7 (Orange) FISH Probe
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
Brand: Abnova™ FC0188.200uL
Additional Details : Weight : 0.02000kg
Specifications
Store at -20°C in the dark. | |
FISH | |
200 μL |