missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12841-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Centromere and kinetochore proteins play a critical role in centromere structure, kinetochore formation, and sister chromatid separation. The protein encoded by this gene colocalizes with inner kinetochore plate proteins CENP-A and CENP-C in both interphase and metaphase. It localizes outside of centromeric heterochromatin, where CENP-B is localized, and inside the kinetochore corona, where CENP-E is localized during prometaphase. It is thought that this protein can bind to itself, as well as to CENP-A, CENP-B or CENP-C. Multimers of the protein localize constitutively to the inner kinetochore plate and play an important role in the organization and function of the active centromere-kinetochore complex.Specifications
CENPH | |
Polyclonal | |
Unconjugated | |
CENPH | |
CENP H, CENPH, centromere protein H, ICEN35, NNF1, PMF1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26886, 64946, 681185 | |
-20°C | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H3R5, Q9QYM8 | |
CENPH | |
CENPH Fusion Protein Ag3836 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |