missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Centri-Sep™ 96-Well Plates
Centri-Sep™ 96's hydrated matrix enables effective and reliable high-throughput removal of excess BigDye™ terminators from DNA sequencing reactions, giving you clean sequencing reads with uniform signal intensity starting with the first base.
2520.00 SEK - 45470.00 SEK
Specifications
Format | 96-well Plate |
---|---|
Product Line | Centri-Sep™ |
Product Type | 96-Well Plate |
Shipping Condition | Wet Ice |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16385302
|
Applied Biosystems™
4367819 |
2 Plates |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16370824
|
Applied Biosystems™
4367821 |
50 Plates |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Processed faster than ultrafiltration columns
- Rapid and efficient separations
- Buffer is not preselected
- Convenient 20 to 100μL sample size
- Dry gel format
- Room Temperature Storage
- Microcentrifuge format
Specifications
96-well Plate | |
Centri-Sep™ | |
96-Well Plate | |
Wet Ice |