Läs mer
Tocris Bioscience™ Centrinone B
High affinity and selective PLK4 inhibitor
Brand: Tocris Bioscience™ 5690/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Centrinone B is a high affinity and selective PLK4 inhibitor (Ki = 0.6 nM). Exhibits >2000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner.Specifikationer
| 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(1-piperidinyl)-4-pyrimidinamine | |
| 1798871-31-4 | |
| LCR 323 | |
| 10 mg |