missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cephalexin Monohydrate USP MP Biomedicals
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
Type | Cephalexin Monohydrate |
Physical Form | Crystalline Powder |
Formula Weight | 365.4 |
Grade | BP/USP |
Color | Beige to White |
Quantity | 5 g |
Assay | ≥95% |
Chemical Name or Material | Cephalexin Monohydrate |
Safety and Handling
Product Identifier
- Cephalexin Monohydrate BP, USP grade
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'