Learn More
Cephradine, MedChemExpress
Product Code. 16468646
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468646 100 mg 100mg
16452256 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16468646 Supplier Medchem Express Supplier No. HYB1156
 Restricted Item
Beräknad leverans: 30-11-2026
MedChemExpress Cephradine (Cefradine) is a broad-spectrum and orally active cephalosporin.
Cephradine is active against both gram-positive and gram-negative pathogens. Cephradine is effective in eradicating most penicillinase-producing organisms. Cephradine has been used in the research of genitourinary, gastrointestinal and respiratory tract infections, and in infections of the skin and soft tissues. Cephradine blocks solar-ultraviolet induced skin inflammation through direct inhibition of TOPK.
Especificaciones
Chemical Name or Material Cephradine
CAS 38821-53-3
Color White
Purity Grade Notes Research
Molecular Formula C16H19N3O4S
Quantity 100 mg
Synonym Cefradine SQ-11436
Solubility Information H2O : 8.33 mg/mL (23.84 mM; Need ultrasonic) ∣DMSO : ≥ 3.6 mg/mL (10.30 mM)
SMILES O=C(C(N12)=C(C)CS[C@]2([H])[C@H](NC([C@H](N)C3=CCC=CC3)=O)C1=O)O
Molecular Weight (g/mol) 349.4
Formula Weight 349.4
Percent Purity 95.11%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Mostrar más Mostrar menos
Research purposes only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.