Läs mer
Ceragenin CSA 44, TRC
Produktkod. 30012584
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012584 50 mg
30011009 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC26230050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1599446-49-7
Molekylformel C41H71N3O8 • 3(HCl)
Molekylvikt (g/mol) 734.0233646
Synonym Octyl (3α,5β,7α,12α)-3,7,12-tris(3-amino-1-oxopropoxy)cholan-24-oate Trihydrochloride, CSA-44 Trihydrochloride;
IUPAC-namn octyl (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-tris(3-aminopropanoyloxy)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate;trihydrochloride
LEDER O=C(CCN)O[C@H]1[C@]([C@@](CC[C@]2([H])[C@H](C)CCC(OCCCCCCCC)=O)([H])[C@]2(C)[C@@H](OC(CCN)=O)C3)([H])[C@@]3([H])[C@]4(C)[C@](C[C@H](OC(CCN)=O)CC4)([H])C1.Cl.Cl.Cl
Specifikationer
Kemiskt namn eller material Ceragenin CSA 44 Trihydrochloride
Kvantitet 50 mg
Frakt skick Room Temperature
InChI formel InChI=1S/C41H71N3O8.3ClH/c1-5-6-7-8-9-10-23-49-35(45)14-11-27(2)30-12-13-31-39-32(26-34(41(30,31)4)52-38(48)18-22-44)40(3)19-15-29(50-36(46)16-20-42)24-28(40)25-33(39)51-37(47)17-21-43;;;/h27-34,39H,5-26,42-44H2,1-4H3;3*1H/t27-,28+,29-,30-,31+,32+,33-,34+,39+,40+,41-;;;/m1.../s1