Learn More
Hamilton™ CERAMIC VALVE, 3-5, MVP, ALUMINUM
The Modular Valve Positioner(MVP) is a self-contained, bi-directional valve positioner used for fluide selection and redirection .
Brand:  Hamilton™ 9412-01
Additional Details : Weight : 0.06400kg
Product Code. 11573444
  • 5010.00 SEK / Each
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
Specification Sheet
Valve Model3-5
Valve Model DescriptionHVC 3-5 MVP
Fluid PathCeramic/PTFE/CTFE
Valve SizeHVC
Nominal Port Diameter0.060 inch (1.5 mm)
Valve Outer Body Dimension0.85 inch (21.6 mm)
Housing MaterialAluminum
Total Dead Volume65 μL per path
ActuatedCeramic Modular Valve Positioner
Break Away Torque15 oz-in (105.9 mNm)
Panel MountingNo
Panel ThicknessN/A
Thread Depth0.320 (8.1 mm)
Threaded Connection¼-28
Operating Temp High35 °C (95 °F)
Operating Temp Low10 °C (50 °F)
Operating Pressure0 to 100 psig (0 to 6.9 bar)
Storage Temp High50 °C (122 °F)
Storage Temp Low-20 °C (-4 °F)
Specifications
Specifications
Ceramic Modular Valve Positioner
Instrument Valve
HVC 3-5 MVP