Learn More
Cerium(IV) trifluoromethanesulfonate, 98%, Thermo Scientific™
473.06 SEK - 1753.58 SEK
Chemical Identifiers
CAS 107792-63-2
Molecular Formula C4CeF12O12S4
Molecular Weight (g/mol) 736.37
MDL Number MFCD03844746
InChI Key BYAJTISDHIASIP-UHFFFAOYSA-J
Synonym cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
PubChem CID 9875257
IUPAC Name cerium(4+);trifluoromethanesulfonate
SMILES [Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11314486
SDS View Documents
Thermo Scientific Alfa Aesar
L20056.06
5g
473.06 SEK
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 6
11324486
SDS View Documents
Thermo Scientific Alfa Aesar
L20056.14
25g
1753.58 SEK
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 5
Description
Description
Applications
Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers
Solubility
Soluble in water.
Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.
Specifications
Chemical Identifiers
107792-63-2
736.37
BYAJTISDHIASIP-UHFFFAOYSA-J
9875257
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
C4CeF12O12S4
MFCD03844746
cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
cerium(4+);trifluoromethanesulfonate
Specifications
Cerium(IV) trifluoromethanesulfonate
5g
C4CeF12O12S4
cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
BYAJTISDHIASIP-UHFFFAOYSA-J
cerium(4+);trifluoromethanesulfonate
9875257
98%
Hygroscopic
107792-63-2
MFCD03844746
Soluble in water.
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
736.37
736.39