Läs mer
Cerivastatin lactone, TRC
Produktkod. 30361349
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30361349 2.5 mg 2.5mg
30360798 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30361349 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2770052.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 158878-47-8
Molekylformel C26H32FNO4
Molekylvikt (g/mol) 441.53
Synonym [4R-[4α,6β(E)]]-6-[2-[4-(4-Fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]ethenyl]tetrahydro-4-hydroxy-2H-pyran-2-one, (4R,6S)-6-((E)-2-(4-(4-Fluorophenyl)-2,6-diisopropyl-5-(methoxymethyl)pyridin-3-yl)vinyl)-4-hydroxytetrahydro-2H-pyran-2-one, (4R,6S)-6-[(1E)-2-[4-(4-Fluorophenyl)-5-(methoxymethyl)-2,6-bis(1-methylethyl)-3-pyridinyl]ethenyl]tetrahydro-4-hydroxy-2H-pyran-2-one
LEDER O=C(C[C@H](O)C1)O[C@@H]1/C=C/C2=C(C3=CC=C(F)C=C3)C(COC)=C(C(C)C)N=C2C(C)C
Specifikationer
Kemiskt namn eller material Cerivastatin lactone
Kvantitet 2.5 mg