missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Chloride, 99.999% (Metals Basis), Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Description
Used for density gradient high speed centrifugation separation of λ phage, M13 and plasmid DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1303°C |
Melting Point | 646°C |
Physical Form | Crystalline Powder |
Quantity | 100 g |
IUPAC Name | caesium(1+) chloride |
Formula Weight | 168.36 |
Percent Purity | 99.999% |
Grade | Molecular Biology |
Packaging | Plastic bottle |
Specific Gravity | 3.99 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'