Läs mer
Cesium Hexafluorophosphate, TRC
Produktkod. 30352135
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30352135 500 mg
30363444 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30352135 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC280025500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16893-41-7
Molekylformel CsF6P
Molekylvikt (g/mol) 277.87
IUPAC-namn cesium;hexafluorophosphate
LEDER [Cs+].F[P-](F)(F)(F)(F)F
Specifikationer
Kemiskt namn eller material Cesium Hexafluorophosphate
Kvantitet 500 mg
InChI formel InChI=1S/Cs.F6P/c;1-7(2,3,4,5)6/q+1;-1
Formel vikt 277.8696