Läs mer
Cetirizine 3-Chloro Impurity Dihydrochloride, TRC
Produktkod. 30133860
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133860 25 mg
17910435 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133860 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36484525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1232460-29-5
Molekylformel C21 H25 Cl N2 O3 . 2 Cl H
Molekylvikt (g/mol) 461.81
Synonym 3-Chlorocetirizine Dihydrochloride, (RS)-2-[2-[4-[(3-Chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic Acid Dihydrochloride, Acetic acid, 2-[2-[4-[(3-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, hydrochloride (1:2) (ACI), [2-[4-[(3-chlorophenyl)phenylmethyl]piperazin-1-yl]ethoxy]acetic acid dihydrochloride
IUPAC-namn 2-[2-[4-[(3-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
LEDER Cl.Cl.OC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3cccc(Cl)c3
Specifikationer
Kemiskt namn eller material 3-Chlorocetirizine Dihydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C21H25ClN2O3.2ClH/c22-19-8-4-7-18(15-19)21(17-5-2-1-3-6-17)24-11-9-23(10-12-24)13-14-27-16-20(25)26;;/h1-8,15,21H,9-14,16H2,(H,25,26);2*1H
Formel vikt 460.1087