missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CETP (Human) Recombinant Protein
Human CETP full-length ORF ( AAH25739.1, 1 a.a. - 493 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001071-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MLAATVLTLALLGNAHACSKGTSHEAGIVCRITKPALLVLNHETAKVIQTAFQRASYPDITGEKAMMLLGQVKYGLHNIQISHLSIASSQVELVEAKSIDVSIQNVSVVFKGTLKYGYTTAWWLGIDQSIDFEIDSAIDLQINTQLTCDSGRVRTDAPDCYLSFHKLLLHLQGEREPGWIKQLFTNFISFTLKLVLKGQICKEINVISNIMADFVQTRAASILSDGDIGVDISLTGDPVITASYLESHHKGHFIYKNVSEDLPLPTFSPTLLGDSRMLYFWFSERVFHSLAKVAFQDGRLMLSLMGDEFKAVLETWGFNTNQEIFQEVVGGFPSQAQVTVHCLKMPKISCQNKGVVVNSSVMVKFLFPRPDQQHSVAYTFEEDIVTTVQASYSKKKLFLSLLDFQITPKTVSNLTESSSESIQSFLQSMITAVGIPEVTSRLEVVFTALMNSKGVSLFDIINPEIITRDGFLLLQMDFGFPEHLLVDFLQSLS
Specifications
AAH25739.1 | |
cholesteryl ester transfer protein, plasma | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CETP | |
GST |
1071 | |
Wheat germ expression system | |
10 μg | |
HDLCQ10 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |