missing translation for 'onlineSavingsMsg'
Learn More

CFHR3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16583-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16896234

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a secreted protein, which belongs to the complement factor H-related protein family. It binds to heparin, and may be involved in complement regulation. Mutations in this gene are associated with decreased risk of age-related macular degeneration, and with an increased risk of atypical hemolytic-uremic syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

CFHR3
Polyclonal
Unconjugated
CFHR3
CFHL3, CFHR3, complement factor H related 3, DOWN16, FHR 3, FHR3, H factor like protein 3, HLF4
Rabbit
Antigen Affinity Chromatography
RUO
10878
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q02985
CFHR3
CFHR3 Fusion Protein Ag9963
20 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers