missing translation for 'onlineSavingsMsg'
Learn More

CFLAR/FLIP Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10394-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876753

  • 4721.67 SEK / 150µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FLIP (CFLAR) is an apoptosis regulator protein which functions as a crucial link between cell survival and cell death pathways in mammalian cells. It acts as an inhibitor of TNFRSF6 mediated apoptosis. A proteolytic fragment (p43) is likely retained in the death-inducing signaling complex (DISC) thereby blocking further recruitment and processing of caspase-8 at the complex. Full length and shorter isoforms have been shown either to induce apoptosis or to reduce TNFRSF-triggered apoptosis. Lacks enzymatic (caspase) activity.
Specifications

Specifications

CFLAR/FLIP
Polyclonal
Unconjugated
CFLAR
c FLIP, c FLIPL, c FLIPR, c FLIPS, CASH, CASP8AP1, Caspase eight related protein, Caspase homolog, Casper, CFLAR, CLARP, FLAME, FLAME 1, FLAME1, FLIP, I FLICE, Inhibitor of FLICE, MRIT, Usurpin
Rabbit
Antigen Affinity Chromatography
RUO
8837
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.72 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15519
CFLAR
CFLAR/FLIP Fusion Protein Ag0593
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers