Läs mer
CFM-2, MedChemExpress
Produktkod. 16475218
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
Förpackningsstorlek:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16475218 10 mg -
16431437 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16475218 Leverantör Medchem Express Leverantörsnummer HY12503
MedChemExpress CFM-2 is a potent and selective non-competitive AMPAR antagonist. CFM-2 possesses anticonvulsant activity in various models of seizures.
CFM-2 is a potent and selective non-competitive AMPAR antagonist. CFM-2 possesses anticonvulsant activity in various models of seizures.
Specifikationer
Kemiskt namn eller material CFM-2
CAS 178616-26-7
Färg Earth Yellow
Anteckningar om renhetsgrad Research
Molekylformel C17H17N3O3
Kvantitet 10 mg
Löslighetsinformation DMSO : ≥ 50 mg/mL (160.60 mM)
LEDER O=C1NN=C(C2=CC=C(N)C=C2)C3=CC(OC)=C(OC)C=C3C1
Molekylvikt (g/mol) 311.34
Formel vikt 311.34
Procent renhet 98.72%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease