missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFTR Mouse, Clone: CFTR/1643, enQuireBio™
Mouse Monoclonal Antibody
2035.00 SEK - 5730.00 SEK
Specifications
Antigen | CFTR |
---|---|
Clone | CFTR/1643 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959757
|
EnquireBio
1080-MSM3-P1 |
100 μg |
5730.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949757
|
EnquireBio
1080-MSM3-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CFTR | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
1080 | |
Recombinant human CFTR fragment (aa258-385) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
165-170kDa |
CFTR/1643 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_011514056 | |
CFTR | |
IgG2b | |
Protein A/G Purified | |
Cell Surface and Cytoplasmic | |
MOLT-4 cells. Pancreas, Kidney or Placenta. |