missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ cGKII Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549822
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The antibody detects endogenous levels of total cGKII protein.
PRKG2 (PKG2, cGKII) exhibits many cellular functions and has cAMP- and cGMP-dependent protein kinase activity. cGKII is thought to play a key role in a diverse set of physiological pathway. cGKII may mediate intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Identification of the pathway that mediates intestinal fluid secretion by E. coli. cGKII is postulated to be a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.
Specifikationer
| cGKII | |
| Polyclonal | |
| Unconjugated | |
| PRKG2 | |
| AW212535; cGK 2; cGK2; cGKII; cGK-II; cGMP dependent protein kinase type II; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; EC 2.7.11.12; GDPKII; PRKG2; PRKGR2; protein kinase cGMP- dependent type 2; protein kinase, cGMP- dependent, type 2; protein kinase, cGMP- dependent, type II; protein kinase, cGMP-dependent, type II; testicular tissue protein Li 99; type II cGMP-dependent protein kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19092, 25523, 5593 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13237, Q61410, Q64595 | |
| PRKG2 | |
| Synthesized peptide derived from internal of human CGK 2 (aa 92-141). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering