Learn More
CGN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531654
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat. Predicted reactivity: Human (97%), Mouse (86%), Rabbit (83%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probably plays a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier. [UniProt]Specifications
CGN | |
Polyclonal | |
Unconjugated | |
CGN | |
6330408J11Rik; CGN; Cingulin; KIAA1319 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9P2M7 | |
CGN | |
Recombinant protein fragment corresponding to a region within amino acids 506 and 722 of Human CGN | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |