Läs mer
Tocris Bioscience™ CGP 36216 hydrochloride
Beskrivning
CGP 36216 hydrochloride is a selective GABAB antagonist. Displays activity at presynaptic receptors; ineffective at GABAB postsynaptic receptors.
Specifikationer
Specifikationer
| CAS | 1781834-71-6 |
| Kvantitet | 10 mg |
| Produkttyp | CGP 36216 hydrochloride |
| Renhet | >99% |
| Molekylvikt (g/mol) | 187.6 |
| Inhibitorer | CGP 36216 hydrochloride |
For Research Use Only