missing translation for 'onlineSavingsMsg'
Läs mer

STEMCELL Technologies CGP77675

Produktkod. 17150332
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17150332 10 mg
17140332 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17150332 Leverantör STEMCELL Technologies Leverantörsnummer 74134

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Src family kinases inhibitor; Inhibits Src, LCK, Yes, Cdc2, EGFR, VEGFR, v-Abl, and FAK

  • CGP77675 is a potent inhibitor of Src family kinases
  • It has been reported to inhibit phosphorylation of peptide substrates and autophosphorylation of purified Src (IC50 = 5 - 20 nM and 40 nM, respectively), Src, and Src kinase family members such as LCK (IC50 = 0.02 mM and 0.29 mM, respectively)
  • It has also been shown to inhibit epidermal growth factor receptor (EGFR), Cdc2, v-Abl, and focal adhesion kinase (FAK) (IC50 = 0.15 mM, > 10 mM, 0.31 mM, and 0.2 mM, respectively; Missbach et al.)
  • In combination with the GSK3 inhibitor CHIR99021 (Catalog #72052), CGP77675 is used to maintain mouse embryonic stem (ES) cells (Shimizu et al.)
  • Preserves the epigenetic and genomic integrity as well as the developmental potential of ES cells (Choi et al.)

Specifikationer

CAS 234772-64-6
Kvantitet 10 mg
Molekylformel C26H29N5O2
Renhet ≥98%
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.