Learn More
CGRP Monoclonal Antibody (5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen ABS0260502
Additional Details : Weight : 0.01000kg
Description
ABS 026-05-02 detects CGRP alpha in human and rat samples. ABS 026-05-02 binds human alpha-CGRP. Based on inhibition ELISA, this antibody shows full cross-reactivity with rat alpha-CGRP and about about 50% cross-reactivity with human beta-CGRP. Cross-reactivity with coated rat alpha-CGRP is low (8%) and there is no cross-reaction with human amylin. ABS 026-05-02 has been successfully used in ELISA and immunohistochemistry procedures.
This gene encodes the peptide hormones calcitonin, calcitonin gene-related peptide and katacalcin by tissue-specific alternative RNA splicing of the gene transcripts and cleavage of inactive precursor proteins. Calcitonin is involved in calcium regulation and acts to regulate phosphorus metabolism. Calcitonin gene-related peptide functions as a vasodilator while katacalcin is a calcium-lowering peptide. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
CGRP | |
Monoclonal | |
1.02 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P01256, P01257, P01258, P06881 | |
Calca | |
Synthetic peptide corresponding to residues 19-37 of human alpha-CGRP. | |
400 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (Paraffin) | |
5 | |
Unconjugated | |
Calca | |
alpha-type CGRP; CA; Cal1; CAL6; CALC; Calc1; Calca; calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6 | |
Mouse | |
Protein A | |
RUO | |
24241, 796 | |
4° C, store in dark | |
Liquid |