Läs mer
Tocris Bioscience™ CGS 19755
GREENER_CHOICE
Produktkod. 18015472
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18015472 10 mg
18096042 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18015472 Leverantör Tocris Bioscience™ Leverantörsnummer 1241/10
Potent competitive NMDA antagonist
Potent, competitive NMDA receptor antagonist. Anticonvulsant and neuroprotective.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110347-85-8
Molekylformel C7H14NO5P
Specifikationer
Kemiskt namn eller material CGS 19755
Kvantitet 10 mg
Formel vikt 223.17
Renhet >98%
Molekylvikt (g/mol) 223.165