Läs mer
Tocris Bioscience™ CGS 35066
:
10mg
Beskrivning
CGS 35066 is a potent endothelin-converting enzyme (ECE) inhibitor that displays > 100-fold selectivity over neutral endopeptidase 24.11 (IC50 values are 22 and 2300 nM respectively). Blocks the hypertensive effects induced by big ET-1 in vitro and reduces the magnitude of cerebral vasospasm following subarachnoid hemorrhage (SAH).
Specifikationer
Specifikationer
| Kvantitet | 10 mg |
| Produkttyp | CGS 35066 |
| Molekylvikt (g/mol) | 349.28 |
| Inhibitorer | CGS 35066 |
For Research Use Only