missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAF1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CHAF1A |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836854
|
Proteintech
17037-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826854
|
Proteintech
17037-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chromatin assembly factor I (CAF1) is a nuclear complex consisting of p50, p60 (CHAF1B), and p150 (CHAF1A) subunits that assembles histone octamers onto replicating DNA in vitro.Specifications
CHAF1A | |
Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q13111, Q9QWF0 | |
10036, 27221 | |
CHAF1A Fusion Protein Ag10536 | |
Primary | |
-20°C | |
CHAF1A |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CAF, CAF 1, CAF 1 subunit A, CAF I 150 kDa subunit, CAF I p150, CAF1, CAF1B, CAF1P150, CHAF1A, hp150, P150 | |
CHAF1A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |