Läs mer
Nabertherm Chamber Furnace LF 1300 Series with B400 Controller
Handla allt Nabertherm Produkter
Kapacitet (metrisk):
15 l
30 l
60 l
120 l
Förpackningsstorlek:
Varje
Beskrivning
- Maximum Temperature: 1300 °C
- Dual shell housing with rear ventilation, provides for low shell temperatures
- Five-sided heating for very good temperature uniformity
- Heating elements on support tubes ensure free heat radiation and a long service life
- Controller mounted on furnace door and removable for comfortable operation
- Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
- High-quality non-classified fiber insulation with corner bricks for shorter heating and cooling times
- Door with brick-on-brick seal, hand fitted
- Short heating times due to high installed power
- Side vent with bypass connection for exhaust pipe
- Self-supporting arch for high stability and greatest possible protection against dust
- Quick lock on door
- Freely adjustable air slide intake in furnace floor
- Stand included
- Defined application within the constraints of the operating instructions
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 15 l |
| Temperatur (metrisk) | 1300°C |
| Temperaturkontroll | B400 |
| Mått (D x B x H) Inredning | 250 x 250 x 250 mm |
| Mått (L x B x H) Exteriör | 860 x 680 x 1230 mm |
| Vikt (metrisk) | 150 kg |