Läs mer
Hanna Instruments™ Champ™ Pocket Sized pH Meter
Område (pH):
0 till 14
Förpackningsstorlek:
Varje
Beskrivning
The series features a rugged and ergonomically enhanced casing that fits comfortably in your hand and a large LCD that makes it easy to read at any angle. The innovation of a non-clogging fiber junction ensures that the meter lasts much longer. Simply pull out 1–2mm (1/8 in.) of the junction fiber to completely renew the junction.
Specifikationer
Specifikationer
| Inkluderar | Protective cap, calibration screwdriver, batteries and instructions |
| Noggrannhet (pH) | ±0.2 |
| Kalibrering | Manual via Trimmers |
| Område (pH) | 0 till 14 |
| Upplösning (pH) | 0.1 |
| Kalibreringspunkter | 1 |
| Visningstyp | LCD |
| Mått (L x B x H) | 175 x 41 x 23 mm (7.9 x 1.8 x 1 in.) |
| Vikt (metrisk) | 78 g |
| Vikt (engelska) | 2.7 oz. |
| Visa mer |