missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Charging Adapters

Available with EU and UK plug

Brand:  Eppendorf™ 4860801006

Additional Details : Weight : 0.18700kg

 View more versions of this product

Product Code. 10470522

  • 520.00 SEK / Each


Estimated Shipment
10-01-2020
Add to basket

Description

Description

  • 230V charging adapter
Specifications

Specifications

230V
1 Each
230 V
EU
Charging Adapter
Videos
Documents