Läs mer
Chartreusin, TRC
Produktkod. 30352374
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30352374 2.5 mg 2.5mg
30368616 1 mg -
30360509 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30352374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2918482.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6377-18-0
Molekylformel C32H32O14
Molekylvikt (g/mol) 640.59
Synonym 10-[[6-Deoxy-2-O-(6-deoxy-3-O-methyl-alpha-D-galactopyranosyl)-beta-D-galactopyranosyl]oxy]-6-hydroxy-1-methylbenzo[h][1]benzopyrano[5,4,3-cde][1]benzopyran-5,12-dione, Antibiotic X 465A, Lambdamycin, NSC 5159
IUPAC-namn 3-[(2S,3R,4S,5R,6R)-3-[(2R,3R,4S,5S,6R)-3,5-dihydroxy-4-methoxy-6-methyloxan-2-yl]oxy-4,5-dihydroxy-6-methyloxan-2-yl]oxy-8-hydroxy-15-methyl-11,18-dioxapentacyclo[10.6.2.02,7.09,19.016,20]icosa-1(19),2(7),3,5,8,12(20),13,15-octaene-10,17-dione
LEDER CO[C@H]1[C@@H](O)[C@@H](C)O[C@H](O[C@H]2[C@H](Oc3cccc4c(O)c5C(=O)Oc6ccc(C)c7C(=O)Oc(c5c67)c34)O[C@H](C)[C@H](O)[C@@H]2O)[C@@H]1O
Specifikationer
Kemiskt namn eller material Chartreusin
Kvantitet 2.5 mg
InChI formel InChI=1S/C32H32O14/c1-10-8-9-15-18-16(10)29(38)45-26-17-13(23(35)20(19(18)26)30(39)43-15)6-5-7-14(17)44-32-28(24(36)21(33)11(2)42-32)46-31-25(37)27(40-4)22(34)12(3)41-31/h5-9,11-12,21-22,24-25,27-28,31-37H,1-4H3/t11-,12-,21+,22+,24+,25-,27+,28-,31-,32+/m1/s1
Formel vikt 640.1792