Learn More
CHD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541424
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: MMRNKDKSQE EDSSLHSNAS SHSASEEASG SDSGSQSESE QGSDPGSGHG Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 90%
CHD2 Polyclonal antibody specifically detects CHD2 in Human samples. It is validated for Western Blot.Specifications
CHD2 | |
Polyclonal | |
Unconjugated | |
CHD2 | |
2810013C04Rik; 2810040A01Rik; 5630401D06Rik; AI851092; ATP-dependent helicase CHD2; BC029703; chd2; CHD-2; chromodomain helicase DNA binding protein 2; chromodomain-helicase-DNA-binding protein 2; DKFZp547I1315; DKFZp686E01200; DKFZp781D1727; Down syndrome cell adhesion molecule; EEOC; FLJ38614; LOW QUALITY PROTEIN: chromodomain-helicase-DNA-binding protein 2; si:ch211-196l7.5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1106 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14647 | |
CHD2 | |
Synthetic peptide directed towards the N-terminal of human CHD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |