Learn More
CHD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13402-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16871594
  • 4721.67 SEK / 150µL
Estimated Shipment: 30-04-2024
to see stock.
Description
Description
The protein encoded by this gene is a 3′-exoribonuclease, with similarity to the RNase D family of 3′-exonucleases. It prefers poly(A) as the substrate, hence, efficiently degrades poly(A) tails of mRNAs. Exonucleolytic degradation of the poly(A) tail is often the first step in the decay of eukaryotic mRNAs. This protein is also involved in silencing of certain maternal mRNAs during oocyte maturation and early embryonic development, as well as in nonsense-mediated decay (NMD) of mRNAs that contain premature stop codons. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CHD9
Polyclonal
Unconjugated
CHD9
AD013, ATP dependent helicase CHD9, CHD 9, CHD9, CReMM, KIAA0308, KISH2, Kismet homolog 2, PRIC320, x0008
Rabbit
Antigen Affinity Chromatography
RUO
109151, 307726, 80205
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3L8U1, Q8BYH8
CHD9
CHD9 Fusion Protein Ag4245
150 μL
Primary
Human, Mouse, Rat