missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHEK2 (CHK2) Recombinant Human Protein

CHEK2 (CHK2) is a serine/threonine kinase and a component of the DNA damage checkpoint pathway.

Brand:  Invitrogen™ PR5460B

Additional Details : Weight : 0.01000kg

Product Code. 15448453

  • 12870.00 SEK / 100µg


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Description

Description

A mutation in the CHK2 gene has been linked to cancer. CHK2 is activated by DNA damage and phosphorylates several modulators of cell cycle control including tumor suppressor proteins.

Specifications

Specifications

NP_009125
Frozen
66kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
RAD53 Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.04% Triton X-100, 4mM DTT, 50% glycerol
100μg
RUO - research use only
CHEK2 (CHK2)
Baculovirus
Human
His Tag
Videos