missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chelerythrine chloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Description
A natural benzophenanthridine alkaloid. Inhibits protein kinase C. Induces apoptosis in leukemia cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A natural benzophenanthridine alkaloid
- Inhibits protein kinase C
- Induces apoptosis in leukemia cells
Specifications
Color | Yellow to Orange |
Quantity | 5 mg |
Merck Index | 14,2051 |
Solubility Information | Soluble in DMSO at 2mg/ml |
IUPAC Name | 1,2-dimethoxy-12-methyl-[1,3]benzodioxolo[5,6-c]phenanthridin-12-ium;chloride |
Formula Weight | 383.82 |
Percent Purity | ≥99% |
Physical Form | Solid |
Chemical Name or Material | Chelerythrine chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'