Learn More
3M™ Chemical Sorbent
Product Code. 10708035
Change view
Click to view available options
Dimensions (L x W):
48c m x 30 m
Unit Size:
Pack of 2
Product Code. Dimensions (L x W) Unit Size
10708035 48c m x 30 m Pack of 2
1 options
Product Code. 10708035 Supplier 3M™ Supplier No. 7000088219
  • ONLINE EXCLUSIVE PRICE
    6925.00 SEK
    3763.12 SEK / Paket med 2
    Save 3161.88 SEK
    45% Av
Beräknad leverans: 09-12-2026
Specifications
Type Chemical Sorbent
Packaging Quantity 2/Cs.
Dimensions (L x W) 48c m x 30 m
Weight (Metric) 8.6 kg