Läs mer
Chenodeoxycholic acid-3-Beta-D-glucuronide, TRC
Produktkod. 30134888
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134888 50 mg
30012039 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC29192050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58814-71-4
Molekylformel C30H48O10
Molekylvikt (g/mol) 568.7
Synonym 24-Norcholane, β-D-glucopyranosiduronic Acid Deriv., Chenodeoxycholic Acid 3-Gucuronide, (3α,5β,7α)-23-Carboxy-7-hydroxy-24-norcholan-3-yl β-D-Glucopyranosiduronic Acid
IUPAC-namn (2S,3S,4S,5R)-6-[[(3R,5R,7R,8R,9S,10S,13R,14S,17R)-17-[(2R)-4-carboxybutan-2-yl]-7-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl]oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER O[C@@H]1C[C@]2([H])C[C@H](O[C@H]3[C@H](O)[C@H](O)[C@H](O)C(C(O)=O)O3)CC[C@]2(C)[C@]4([H])[C@]1([H])[C@@]5([H])[C@]([C@]([C@H](C)CCC(O)=O)([H])CC5)(C)CC4
Specifikationer
Kemiskt namn eller material Chenodeoxycholic Acid-3-beta-D-glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C30H48O10/c1-14(4-7-21(32)33)17-5-6-18-22-19(9-11-30(17,18)3)29(2)10-8-16(12-15(29)13-20(22)31)39-28-25(36)23(34)24(35)26(40-28)27(37)38/h14-20,22-26,28,31,34-36H,4-13H2,1-3H3,(H,32,33)(H,37,38)/t14-,15+,16-,17-,18+,19+,20-,22+,23+,24+,25-,26+,28?,29+,30-/m1/s1
Procent renhet >95%