Läs mer
Chenopodium Oil, TRC
Produktkod. 30361227
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30361227 1 g
1 options
Produktkod. 30361227 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC2919301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chenopodium Oil
CAS 8006-99-3
Kvantitet 1 g
Synonym Oil Of American Wormseed, Oil Of Chenopodium