missing translation for 'onlineSavingsMsg'
Learn More

CHES, 0.5M buffer soln, pH 8.5, Thermo Scientific™

Catalog Number p-7042150
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
C8H17NO3S
207.288
MFCD00003835
MKWKNSIESPFAQN-UHFFFAOYSA-N
66898
2-(cyclohexylamino)ethanesulfonic acid
C1CCC(CC1)NCCS(=O)(=O)O
This item is not returnable. View return policy
C8H17NO3S
207.288
MFCD00003835
MKWKNSIESPFAQN-UHFFFAOYSA-N
66898
2-(cyclohexylamino)ethanesulfonic acid
C1CCC(CC1)NCCS(=O)(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Concentration 0.5 M
pH 8.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material CHES buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'