missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CHES1 (Human) Recombinant Protein
Human CHES1 full-length ORF ( AAH07506, 1 a.a. - 468 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001112-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGPVMPPSKKPESSGISVSSGLSQCYGGSGFSKALQEDDDLDFSLPDIRLEEGAMEDEELTNLNWLHESKNLLKSFGESVLRSVSPVQDLDDDTPPSPAHSDMPYDARQNPNCKPPYSFSCLIFMAIEDSPTKRLPVKDIYNWILEHFPYFANAPTGWKNSVRHNLSLNKCFKKVDKERSQSIGKGSLWCIDPEYRQNLIQALKKTPYHPHPHVFNTPPTCPQAYQSTSGPPIWPGSTFFKRNGALLQVPPGVIQNGARVLSRGLFPGVRPLPITPIGVTAAMRNGITSCRMRTESEPSCGSPVVSGDPKEDHNYSSAKSSNARSTSPTSDSISSSSSSADDHYEFATKGSQEGSEGSEGSFRSHESPSDTEEDDRKHSQKEPKDSLGDSGYASQHKKRQHFAKARKVPSDTLPLKKRRTEKPPESDDEEMKEAAGSLLHLAGIRSCLNNITNRTAKGQKEQKETTKN
Specifications
AAH07506 | |
forkhead box N3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FOXN3 | |
GST |
1112 | |
Wheat germ expression system | |
25 μg | |
C14orf116, CHES1, PRO1635 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |