Läs mer
Chicoric Acid, TRC
Produktkod. 30361770
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30361770 25 mg
1 options
Produktkod. 30361770 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC29197025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cichoric Acid
CAS 6537-80-0
Anteckningar om renhetsgrad HPLC
Molekylformel C22 H18 O12
Kvantitet 25 mg
Synonym Butanedioic acid, 2,3-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-, (2R,3R)- (9CI), Butanedioic acid, 2,3-bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-, [R-(R*,R*)]-, Tartaric acid, bis(3,4-dihydroxycinnamate) (6CI,8CI), (2R,3R)-2,3-Bis[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]butanedioic acid, Chicoric acid, Cichoric acid, Dicaffeoyltartaric acid
InChI formel InChI=1S/C22H18O12/c23-13-5-1-11(9-15(13)25)3-7-17(27)33-19(21(29)30)20(22(31)32)34-18(28)8-4-12-2-6-14(24)16(26)10-12/h1-10,19-20,23-26H,(H,29,30)(H,31,32)/t19-,20-/m1/s1
LEDER OC(=O)[C@H](OC(=O)C=Cc1ccc(O)c(O)c1)[C@@H](OC(=O)C=Cc2ccc(O)c(O)c2)C(=O)O
IUPAC-namn (2R,3R)-2,3-bis[3-(3,4-dihydroxyphenyl)prop-2-enoyloxy]butanedioic acid
Molekylvikt (g/mol) 474.37
Formel vikt 474.0798