Läs mer
CHIR-99021 Hydrochloride, TRC
Produktkod. 30360289
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30360289 10 mg
1 options
Produktkod. 30360289 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC29215010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-[(2-[(4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl)amino]ethyl)amino]-3-pyridinecarbonitrile Hydrochloride
CAS 1797989-42-4
Molekylformel C22H19Cl3N8
Kvantitet 10 mg
Synonym CHIR-99021 Hydrochloride, CHIR 911 Hydrochloride, CT 99021 Hydrochloride, CHIR99021
InChI formel InChI=1S/C22H18Cl2N8.ClH/c1-13-10-29-21(31-13)17-12-30-22(32-20(17)16-4-3-15(23)8-18(16)24)27-7-6-26-19-5-2-14(9-25)11-28-19;/h2-5,8,10-12H,6-7H2,1H3,(H,26,28)(H,29,31)(H,27,30,32);1H
LEDER Cl.Cc1c[nH]c(n1)c2cnc(NCCNc3ccc(cn3)C#N)nc2c4ccc(Cl)cc4Cl
IUPAC-namn 6-[2-[[4-(2,4-dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)pyrimidin-2-yl]amino]ethylamino]pyridine-3-carbonitrile;hydrochloride
Molekylvikt (g/mol) 501.8