Läs mer
Chlonixin, TRC
Produktkod. 17904715
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17904715 500 mg
17942035 100 mg
17904655 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17904715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC587140500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17737-65-4
Molekylformel C13 H11 Cl N2 O2
Molekylvikt (g/mol) 262.69
Synonym 2-[(3-Chloro-2-methylphenyl)amino]-3-pyridinecarboxylic Acid;
IUPAC-namn 2-(3-chloro-2-methylanilino)pyridine-3-carboxylic acid
LEDER Cc1c(Cl)cccc1Nc2ncccc2C(=O)O
Specifikationer
Kemiskt namn eller material Chlonixin
Kvantitet 500 mg
InChI formel InChI=1S/C13H11ClN2O2/c1-8-10(14)5-2-6-11(8)16-12-9(13(17)18)4-3-7-15-12/h2-7H,1H3,(H,15,16)(H,17,18)
Formel vikt 262.0509