Läs mer
Chlor Cyclizine Hydrochloride, TRC
Produktkod. 30367019
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367019 100 mg
30358259 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367019 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC326500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14362-31-3
Molekylformel C18 H21 Cl N2 . Cl H
Molekylvikt (g/mol) 337.29
Synonym Chlorcyclizine Hydrochloride, Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-methyl-, hydrochloride (1:1), Piperazine, 1-(p-chloro-α-phenylbenzyl)-4-methyl-, monohydrochloride (8CI), Piperazine, 1-[(4-chlorophenyl)phenylmethyl]-4-methyl-, monohydrochloride (9CI), Chlorcyclizine chloride, Chlorcyclizine hydrochloride, Chlorcyclizine monohydrochloride, Di-Paralene, Histantin, NSC 169496, Perazil
IUPAC-namn 1-[(4-chlorophenyl)-phenylmethyl]-4-methylpiperazine;hydrochloride
LEDER Cl.CN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
Specifikationer
Kemiskt namn eller material Chlorcyclizine Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C18H21ClN2.ClH/c1-20-11-13-21(14-12-20)18(15-5-3-2-4-6-15)16-7-9-17(19)10-8-16;/h2-10,18H,11-14H2,1H3;1H
Formel vikt 336.116