Läs mer
Chloramphenicol 1-Acetate, TRC
Produktkod. 30349097
Change view
Kvantitet:
1 mg
2.5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349097 25 mg
30356249 2.5 mg
30366762 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349097 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC32509025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23214-93-9
Molekylformel C13H14Cl2N2O6
Molekylvikt (g/mol) 365.17
Synonym N-[(1R,2R)-2-(Acetyloxy)-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-2,2-dichloro-acetamide, 1-Acetoxychloramphenicol, 1-Acetylchloramphenicol, 1-O-Acetylchloramphenicol, Chloramphenicol 1-Acetate, Chloramphenicol 1'-Acetate
IUPAC-namn [(1R,2R)-2-[(2,2-dichloroacetyl)amino]-3-hydroxy-1-(4-nitrophenyl)propyl] acetate
LEDER CC(=O)O[C@H](C1=CC=C(C=C1)[N+](=O)[O-])[C@@H](CO)NC(=O)C(Cl)Cl
Specifikationer
Kemiskt namn eller material Chloramphenicol 1-Acetate
Kvantitet 25 mg
InChI formel InChI=1S/C13H14Cl2N2O6/c1-7(19)23-11(10(6-18)16-13(20)12(14)15)8-2-4-9(5-3-8)17(21)22/h2-5,10-12,18H,6H2,1H3,(H,16,20)/t10-,11-/m1/s1
Formel vikt 364.0229