Läs mer
Chlorbufam, TRC
Produktkod. 30361402
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30361402 250 mg
1 options
Produktkod. 30361402 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC292840250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chlorbufam
CAS 1967-16-4
Molekylformel C11 H10 Cl N O2
Kvantitet 250 mg
Synonym Carbamic acid, (3-chlorophenyl)-, 1-methyl-2-propynyl ester (9CI), Carbanilic acid, m-chloro-, 1-methyl-2-propynyl ester (6CI,8CI), 1-Butyn-3-yl m-chlorophenylcarbamate, 3-Butyn-2-yl m-chlorocarbanilate, 3-Chlorophenylcarbamic acid 1-methylpropynyl ester, BIPC, Chlorbufam, Chlorbupham, Chlorobufam, Grisemin
InChI formel InChI=1S/C11H10ClNO2/c1-3-8(2)15-11(14)13-10-6-4-5-9(12)7-10/h1,4-8H,2H3,(H,13,14)
LEDER CC(OC(=O)Nc1cccc(Cl)c1)C#C
IUPAC-namn but-3-yn-2-yl N-(3-chlorophenyl)carbamate
Molekylvikt (g/mol) 223.66
Formel vikt 223.04