Läs mer
Chlordimeform, TRC
Produktkod. 30360660
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30360660 1 g
1 options
Produktkod. 30360660 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3270601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Chlordimeform free base
CAS 6164-98-3
Molekylformel C10 H13 Cl N2
Kvantitet 1 g
Synonym Formamidine, N'-(4-chloro-o-tolyl)-N,N-dimethyl- (7CI,8CI), N'-(4-Chloro-2-methylphenyl)-N,N-dimethylmethanimidamide, 1-(2-Methyl-4-chlorophenyl)-3,3-dimethylformamidine, Acaron, C 8514, CDM, CDM (acaricide), CIBA 8514, Chlordimeform, Chlorophenamidin, Chlorophenamidine, Chlorphenamidine, Formamidine, N,N-dimethyl-N'-(2-methyl-4-chlorophenyl)-, Fundal, Fundal 500, Galecron, N,N-Dimethyl-N'-(2-methyl-4-chlorophenyl)formamidine, N'-(2-Methyl-4-chlorophenyl)-N,N-dimethylformamidine, N'-(4-Chloro-o-tolyl)-N,N-dimethylformamidine, N'-(p-Chloro-o-tolyl)-N,N-dimethylformamidine, RS 141, Schering 36,268
InChI formel InChI=1S/C10H13ClN2/c1-8-6-9(11)4-5-10(8)12-7-13(2)3/h4-7H,1-3H3
LEDER CN(C)C=Nc1ccc(Cl)cc1C
IUPAC-namn N'-(4-chloro-2-methylphenyl)-N,N-dimethylmethanimidamide
Molekylvikt (g/mol) 196.68
Formel vikt 196.0767