Läs mer
Chlorhexidine Diacetate Impurity A, TRC
Produktkod. 30012658
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012658 5 mg
30009006 10 mg
30011772 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012658 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3349955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 152504-08-0
Molekylformel C16 H24 Cl N9
Molekylvikt (g/mol) 377.88
Synonym Chlorhexidine nitrile, 1-(4-Chlorophenyl)-5-[6-[(cyanocarbamimidoyl)amino]hexyl]biguanide, Imidodicarbonimidic diamide, N-(4-chlorophenyl)-N'-[6-[[(cyanoamino)iminomethyl]amino]hexyl]-, N-(4-Chlorophenyl)-N'-[6-[[(cyanoamino)iminomethyl]amino]hexyl]imidodicarbonimidic diamide
IUPAC-namn 1-(4-chlorophenyl)-3-[N-[6-[(N-cyanocarbamimidoyl)amino]hexyl]carbamimidoyl]guanidine
LEDER Clc1ccc(NC(=N)NC(=N)NCCCCCCNC(=N)NC#N)cc1
Specifikationer
Kemiskt namn eller material Chlorhexidine nitrile
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C16H24ClN9/c17-12-5-7-13(8-6-12)25-16(21)26-15(20)23-10-4-2-1-3-9-22-14(19)24-11-18/h5-8H,1-4,9-10H2,(H3,19,22,24)(H5,20,21,23,25,26)
Formel vikt 377.1843
Procent renhet >95%